Learn More
Medchemexpress LLC BX517 | 850717-64-5 | 98.0% | 282.30 | 100 MG
SDP
Catalog No. 5000351771
Encompass_Preferred
BX517 is a potent and selective inhibitor of PDK1, demonstrating an IC50 of 6 nM. It effectively blocks the activation of Akt in various tumor cells with submicromolar potency. This compound exhibits significant selectivity, being at least 100-fold more selective against PDK1 compared to a panel of seven other Ser/Thr and Tyr kinases.
  • Potent and selective inhibitor of PDK1
  • Blocks activation of Akt in tumor cells
  • Exhibits submicromolar potency
  • Highly selective against other Ser/Thr and Tyr kinases
Catalog No. 50-003-51771 Supplier Medchemexpress LLC Supplier No. HY13842100MG
BX517 is a potent and selective inhibitor of PDK1, demonstrating an IC50 of 6 nM. It effectively blocks the activation of Akt in various tumor cells with submicromolar potency. This compound exhibits significant selectivity, being at least 100-fold more selective against PDK1 compared to a panel of seven other Ser/Thr and Tyr kinases.
  • Potent and selective inhibitor of PDK1
  • Blocks activation of Akt in tumor cells
  • Exhibits submicromolar potency
  • Highly selective against other Ser/Thr and Tyr kinases