Learn More
Enzo Life Sciences BX795 (10mg). CAS: 702675-74-9
SDP
Catalog No. 502013503
Encompass_Preferred
BX795 was initially developed as a PDK1 inhibitor (direct competitive inhibitor IC50 = 11nM for PDK1) and has been shown to be a potent and relatively specific inhibitor of TBK1 and IKK-ε. It blocks the phosphorylation, nuclear translocation, and transcriptional activity of interferon regulatory factor 3. BX795 also is a potent inhibitor of cell growth of multiple cancer cell lines with IC50 values ranging from submicromolar amounts (0.368 µM to greater than 450 µM). Alternative name: N-[3-[[5-iodo-4-[3-(thiophene-2-carbonylamino)propylamino]pyrimidin-2-yl]amino]phenyl]pyrrolidine-1-carboxamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (118mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3503 Supplier Enzo Life Sciences Supplier No. ENZCHM1890010
BX795 was initially developed as a PDK1 inhibitor (direct competitive inhibitor IC50 = 11nM for PDK1) and has been shown to be a potent and relatively specific inhibitor of TBK1 and IKK-ε. It blocks the phosphorylation, nuclear translocation, and transcriptional activity of interferon regulatory factor 3. BX795 also is a potent inhibitor of cell growth of multiple cancer cell lines with IC50 values ranging from submicromolar amounts (0.368 µM to greater than 450 µM). Alternative name: N-[3-[[5-iodo-4-[3-(thiophene-2-carbonylamino)propylamino]pyrimidin-2-yl]amino]phenyl]pyrrolidine-1-carboxamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (118mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.