Learn More
Aviva Systems Biology Corp BXDC5 Antibody - N-terminal region (ARP41019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BXDC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BXDC5. Alternate names: DKFZp761G0415, DKFZp761M0215, FLJ12475, RP11-118B23.1, RPF1, BXDC5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-53261