Learn More
Cayman Chemical ByakngelIcIn 10mg
SDP
Catalog No. 502948552
Encompass_Preferred
A coumarin; inhibits the activity of aldose reductase (IC50 = 6.2 µM for the bovine lens enzyme) and reduces cataract formation in a rat model of galactosemia at 50 mg/kg per day; decreases LPS-induced PGE2 production in isolated rat peritoneal macrophages at 30 µM; reduces cartilage erosion and loss of proteoglycan in the inflamed joints of a mouse model of surgically induced osteoarthritis at 10 mg/kg
Catalog No. 50-294-8552 Supplier Cayman Chemical Supplier No. 3460010 MG
A coumarin; inhibits the activity of aldose reductase (IC50 = 6.2 µM for the bovine lens enzyme) and reduces cataract formation in a rat model of galactosemia at 50 mg/kg per day; decreases LPS-induced PGE2 production in isolated rat peritoneal macrophages at 30 µM; reduces cartilage erosion and loss of proteoglycan in the inflamed joints of a mouse model of surgically induced osteoarthritis at 10 mg/kg