Learn More
Cayman Chemical Byakangelicol, Purity: ≥98%, CAS: 26091-79-2, 10 mg
SDP
Catalog No. 502948801
Encompass_Preferred
A furanocoumarin with diverse biological activities; inhibits AChE and BChE (IC50s = 46.3 and 45.2 µM, respectively); inhibits LPS-induced production of NO in RAW 264.7 cells (IC50 = 16.9 µg/ml) and IL-1β-induced COX-2 activity and PGE2 release in A549 cells from 10-50 µM; reduces tacrine-induced cytotoxicity in HepG2 cells (EC50 = 36.6 µM)
Catalog No. 50-294-8801 Supplier Cayman Chemical Supplier No. 3457110 MG
A furanocoumarin with diverse biological activities; inhibits AChE and BChE (IC50s = 46.3 and 45.2 µM, respectively); inhibits LPS-induced production of NO in RAW 264.7 cells (IC50 = 16.9 µg/ml) and IL-1β-induced COX-2 activity and PGE2 release in A549 cells from 10-50 µM; reduces tacrine-induced cytotoxicity in HepG2 cells (EC50 = 36.6 µM)