Learn More
TARGETMOL CHEMICALS INC BYK204165 25MG
SDP
Catalog No. 502579108
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BYK204165 (RT-017290) is a potent PARP inhibitor (PARP1; IC50 = 44.67 nM for human recombinant PARP1 in an enzyme assay)purity: 99%
Catalog No. 50-257-9108 Supplier TARGETMOL CHEMICALS INC Supplier No. T789625MG
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BYK204165 (RT-017290) is a potent PARP inhibitor (PARP1; IC50 = 44.67 nM for human recombinant PARP1 in an enzyme assay)purity: 99%