Learn More
Apexbio Technology LLC BYL-719 1217486-61-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062215
Encompass_Preferred
BYL-719 (CAS 1217486-61-7) is a selective inhibitor of PI3K demonstrating minimal activity against PI3K or isoforms Through targeting PI3K an essential node frequently dysregulated via PIK3CA mutations in various cancers BYL-719 effectively disrupts downstream proliferative signaling In biochemical assays BYL-719 shows potent inhibition of PI3K with an IC50 of 5 nM Preclinical evaluations indicate dose-dependent antitumor efficacy in xenograft tumor models and proliferation arrest and apoptosis induction in multiple myeloma cell lines exhibiting PIK3CA overexpression Clinically initial studies reveal manageable safety profiles and preliminary antitumor responses notably in PIK3CA-mutant malignancies supporting continued exploration in oncology research settings
Catalog No. 50-000-62215 Supplier Apexbio Technology LLC Supplier No. A83465.1
BYL-719 (CAS 1217486-61-7) is a selective inhibitor of PI3K demonstrating minimal activity against PI3K or isoforms Through targeting PI3K an essential node frequently dysregulated via PIK3CA mutations in various cancers BYL-719 effectively disrupts downstream proliferative signaling In biochemical assays BYL-719 shows potent inhibition of PI3K with an IC50 of 5 nM Preclinical evaluations indicate dose-dependent antitumor efficacy in xenograft tumor models and proliferation arrest and apoptosis induction in multiple myeloma cell lines exhibiting PIK3CA overexpression Clinically initial studies reveal manageable safety profiles and preliminary antitumor responses notably in PIK3CA-mutant malignancies supporting continued exploration in oncology research settings