Learn More
Apexbio Technology LLC BYL-719, 5mg. Cas: 1217486-61-7 MFCD: MFCD22417085. Selective PI3Kα inhibitor
SDP
Supplier:  Apexbio Technology LLC A83465
Encompass_Preferred
BYL719 is a selective PI3K inhibitor with IC50 of 5 nM. It has minimal effect on PI3K, and [1]. Dysregulation of the PI3K signaling pathway is involved in multiple cancers. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5123