Learn More
Selleck Chemical LLC Alpelisib (BYL719) 5mg 1217486-61-7
SDP
Supplier:  Selleck Chemical LLC S28145MG
Encompass_Preferred
Alpelisib (BYL719) is a potent and selective PI3K? inhibitor with IC50 of 5 nM in a cell-free assay, and minimal effect on PI3K?/?/?. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0421898