Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bz-Ala-Arg TFA | 99.8% | 463.41 | 10 MG
SDP

Catalog No. 5000400061
Encompass_Preferred

Bz-Ala-Arg TFA is a dipeptide that functions as a substrate for human pancreatic carboxypeptidase B and plasma carboxypeptidase N.

  • Functions as a dipeptide
  • Substrate for human pancreatic carboxypeptidase B
  • Substrate for plasma carboxypeptidase N
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Requires sealed storage, away from moisture and light

Catalog No. 50-004-00061 Supplier Medchemexpress LLC Supplier No. HYP4522A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bz-Ala-Arg TFA is a dipeptide that functions as a substrate for human pancreatic carboxypeptidase B and plasma carboxypeptidase N.

  • Functions as a dipeptide
  • Substrate for human pancreatic carboxypeptidase B
  • Substrate for plasma carboxypeptidase N
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Requires sealed storage, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.