Learn More
Carbosynth LLC. Bz-Arg-NH2 HCl H20, 5g, MW: 277.32 36.46 18.02, Molecular formula: C13H19N5O2 HCI H2O, CAS# 4299-03-0
SDP
Supplier:  Carbosynth LLC. SRA30025G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Benzoyl-L-Arginine Amide Monohydrochloride Monohydrate, Purity: , Grade AA, Application: Substrate for Trypsin, Storage Temperature: -20C, References: K.A. Walsh, In, Proteolytic Enzymes, Methods in Enzymology, Vol. 19, (G.E. Perlmann and L. Lorand, Eds.), Academic Press, New York, 1970, pp. 41-63.
Catalog No. 50-168-7149