Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bz-dA (N6-Benzoyl-2'-deoxyadenosine) | 4546-72-9 | 98.4% | 355.35 | 25 G
SDP

Catalog No. 5000366891
Encompass_Preferred

Bz-dA (N6-Benzoyl-2'-deoxyadenosine) is a nucleoside analog primarily used for the synthesis of oligoribonucleotides. It is supplied as a white to off-white solid with a high purity of 98.43%. This product is intended for research purposes only.

  • Nucleoside analog for oligoribonucleotide synthesis.
  • Purity is 98.43%.
  • Molecular weight is 355.35.
  • Appearance is a white to off-white solid.
  • Recommended storage for powder is -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months, -20°C for 1 month.
  • Highly soluble in DMSO.
  • For research use only.

Catalog No. 50-003-66891 Supplier Medchemexpress LLC Supplier No. 25GHYW011125
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bz-dA (N6-Benzoyl-2'-deoxyadenosine) is a nucleoside analog primarily used for the synthesis of oligoribonucleotides. It is supplied as a white to off-white solid with a high purity of 98.43%. This product is intended for research purposes only.

  • Nucleoside analog for oligoribonucleotide synthesis.
  • Purity is 98.43%.
  • Molecular weight is 355.35.
  • Appearance is a white to off-white solid.
  • Recommended storage for powder is -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months, -20°C for 1 month.
  • Highly soluble in DMSO.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.