Learn More
Bachem Bz-DL-Arg-pNA · HCl
SDP
Supplier:  Bachem 4001028.0005
Encompass_Preferred
Bz-DL-Arg-pNA · HCl, 911-77-3, C19H22N6O4 · HCl, Mr 434.88, 5g. DL-BAPA · HCl, DL-BApNA · HCl. Substrate for trypsin, papain, acrosin, plasmin, cathepsin B, and cathepsin H.
Catalog No. 50-396-898