Learn More
Medchemexpress LLC Bz-DTPA | 102650-30-6 | 96.1% | 540.54 g/mol | C22H28N4O10S | 10 MG
SDP
Catalog No. 5000287719
Encompass_Preferred
Bz-DTPA is a bifunctional chelator used to conjugate metal radioisotopes to biomolecules such as monoclonal antibodies. It is commonly employed in research to prepare immunoconjugates for indium-111 labeling. The product is supplied as a solid and is also available as a DMSO solution for convenience.
  • Bifunctional chelator suitable for radiolabeling biomolecules.
  • Used to prepare immunoconjugates for indium-111 labeling.
  • Reported purity approximately 96.05%.
  • Available as solid and as a 10 mM solution in DMSO.
  • Available in small pack sizes (e.g., 10 mg) for research workflows.
  • Store solid at 4°C protected from light; store solutions at -80°C or -20°C per guidance.
Catalog No. 50-002-87719 Supplier Medchemexpress LLC Supplier No. HY15603510MG
Bz-DTPA is a bifunctional chelator used to conjugate metal radioisotopes to biomolecules such as monoclonal antibodies. It is commonly employed in research to prepare immunoconjugates for indium-111 labeling. The product is supplied as a solid and is also available as a DMSO solution for convenience.
  • Bifunctional chelator suitable for radiolabeling biomolecules.
  • Used to prepare immunoconjugates for indium-111 labeling.
  • Reported purity approximately 96.05%.
  • Available as solid and as a 10 mM solution in DMSO.
  • Available in small pack sizes (e.g., 10 mg) for research workflows.
  • Store solid at 4°C protected from light; store solutions at -80°C or -20°C per guidance.