Learn More
Carbosynth LLC. Bz-Gly-His-Leu H2O [Hippuryl-Histidyl-Leucine], 0.1g, MW: 429.47 18.02, Molecular formula: C21H27N5O5 H2O, CAS# 31373-65-6
SDP
Supplier:  Carbosynth LLC. SGL30640.1G
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzoyl-Glycyl-L-Histidyl-L-Leucine, Purity: , Grade AA, Application: Substrate for ACE (Angiotensin I Converting Enzyme), Storage Temperature: -20C, References: D.W. Cushman and H.S. Cheung, Biochem. Pharmacol., 20, 1637 (1971).
Catalog No. 50-168-7103