Learn More
Cayman Chemical Bz-IEGR-pNAacetate 10mg
SDP
Catalog No. 502941681
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-294-1681 Supplier Cayman Chemical Supplier No. 2756710 MG
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity