Learn More
Cayman Chemical Bz-IEGR-pNAacetate 1mg
SDP
Catalog No. 502940254
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-294-0254 Supplier Cayman Chemical Supplier No. 275671 MG
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity