Learn More
Cayman Chemical Bz-IEGR-pNA (acetate), Purity: ≥98%, CAS: 1325307-57-0, 25 mg
SDP
Catalog No. 502939463
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-293-9463 Supplier Cayman Chemical Supplier No. 2756725 MG
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity