Learn More
Cayman Chemical Bz-IEGR-pNAacetate 5mg
SDP
Catalog No. 502937739
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-293-7739 Supplier Cayman Chemical Supplier No. 275675 MG
A colorimetric Factor Xa substrate; cleavage of the IEGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity