Learn More
Carbosynth LLC. Bz-L-Arg-pNA • HCl, 5g, MW: 398.42 • 36.46, Molecular formula: C19H22N6O4 • HCI, CAS# 21653-40-7
SDP
Supplier:  Carbosynth LLC. SRN30575G
Encompass_Preferred
Synonyms: [L-BAPA] Benzoyl-L-Arginine p-Nitroanilide Monohydrochloride, Purity: , Grade AA, Application: Substrate for Trypsin-like Proteases and Papain, Storage Temperature: -20°C, References: K.A. Walsh, In, Proteolytic Enzymes, Methods in Enzymology, Vol. 19, (G.E. Perlmann and L. Lorand, Eds.), Academic Press, New York, 1970, pp. 41-63., R. Arnon, In, Proteolytic Enzymes, Methods in Enzymology, Vol. 19, (G.E. Perlmann and L. Lorand, Eds.), Academic Press, New York, 1970, pp. 226-244., G.A. Grant, A.Z. Eisen, and R.A. Bradshaw, In, Proteolytic Enzymes Part C, Methods in Enzymology, Vol. 80, (L. Lorand, ed.), Academic Press, New York, 1981, pp. 722-734.
Catalog No. 50-168-7163