Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bz-(Me)Tz-NHS | 1454558-58-7 | C19H20N6O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1516865MG

Encompass_Preferred

Bz-(Me)Tz-NHS is a click chemistry reagent containing a methyltetrazine group. It is a 3rd generation Click-Linker designed for Cu-free click conjugation.

  • Click chemistry reagent
  • Contains a methyltetrazine group
  • 3rd generation Click-Linker for Cu-free conjugation
  • Purity of 95.02%
  • Molecular weight of 412.40
  • Appears as a purple to purplish red solid
  • Soluble in DMSO at 25 mg/mL (60.62 mM)

Catalog No. 50-003-73436


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.