Learn More
Medchemexpress LLC Bz-(Me)Tz-NHS | 1454558-58-7 | MFCD00076258 | C₁₉H₂₀N₅O₅S | 25 MG
SDP
Catalog No. 5000374250
Encompass_Preferred
Bz-(Me)Tz-NHS is a click chemistry reagent that contains a methyltetrazine group. It functions as a 3rd generation Click-Linker for Cu-free click conjugation, primarily used in antibody-drug conjugate (ADC) related research.
  • Contains a methyltetrazine group
  • Functions as a 3rd generation Click-Linker
  • Designed for Cu-free click conjugation
  • Used in antibody-drug conjugate (ADC) related research
Catalog No. 50-003-74250 Supplier Medchemexpress LLC Supplier No. 25MGHY151686
Bz-(Me)Tz-NHS is a click chemistry reagent that contains a methyltetrazine group. It functions as a 3rd generation Click-Linker for Cu-free click conjugation, primarily used in antibody-drug conjugate (ADC) related research.
  • Contains a methyltetrazine group
  • Functions as a 3rd generation Click-Linker
  • Designed for Cu-free click conjugation
  • Used in antibody-drug conjugate (ADC) related research