Learn More
Bachem Bz-Phe-Val-Arg-AMC Hydrochloride
SDP
Catalog No. 50260275
Encompass_Preferred
Bz-Phe-Val-Arg-AMC Hydrochloride, 88899-22-3, C37H43N7O6, Mr 681.79, 25mg. Thrombin Substrate III, Fluorogenic. Sensitive fluorogenic substrate for the quantitative determination of thrombin. Bz-FVR-AMC is also cleaved by ficain.
Catalog No. 50-260-275 Supplier Bachem Supplier No. 4003131.0025
Add to Cart
Add to Cart
Bz-Phe-Val-Arg-AMC Hydrochloride, 88899-22-3, C37H43N7O6, Mr 681.79, 25mg. Thrombin Substrate III, Fluorogenic. Sensitive fluorogenic substrate for the quantitative determination of thrombin. Bz-FVR-AMC is also cleaved by ficain.