Learn More
Medchemexpress LLC Bz-Phe-Val-Arg-AMC | 88899-22-3 | 99.8% | 681.78 g·mol⁻¹ | C37H43N7O6 | 10 MG
SDP
Catalog No. 5000297086
Encompass_Preferred
Bz-FVR-AMC is a fluorogenic peptide substrate used to monitor procathepsin and related protease activity. The substrate releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage and has an observed kcat/Km of 1,070 mM⁻¹·s⁻¹; at high concentrations it can inhibit the reaction. Typical handling: supplied as a solid and stored frozen to maintain stability.
  • Releases the fluorescent AMC reporter upon enzymatic cleavage.
  • Used to measure procathepsin and related protease activity in biochemical assays.
  • Reported kcat/Km of 1,070 mM⁻¹·s⁻¹.
  • High concentrations may inhibit protease activity.
  • Supplied as a solid and stored at -20 °C to maintain stability.
  • Reported purity about 99.8% and molecular weight 681.78 g·mol⁻¹.
Catalog No. 50-002-97086 Supplier Medchemexpress LLC Supplier No. HYD163410MG
Bz-FVR-AMC is a fluorogenic peptide substrate used to monitor procathepsin and related protease activity. The substrate releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage and has an observed kcat/Km of 1,070 mM⁻¹·s⁻¹; at high concentrations it can inhibit the reaction. Typical handling: supplied as a solid and stored frozen to maintain stability.
  • Releases the fluorescent AMC reporter upon enzymatic cleavage.
  • Used to measure procathepsin and related protease activity in biochemical assays.
  • Reported kcat/Km of 1,070 mM⁻¹·s⁻¹.
  • High concentrations may inhibit protease activity.
  • Supplied as a solid and stored at -20 °C to maintain stability.
  • Reported purity about 99.8% and molecular weight 681.78 g·mol⁻¹.