Learn More
Medchemexpress LLC Bz-rA Phosphoramidite | 104992-55-4 | 988.19 | 1 ML
SDP
Catalog No. 5000446600
Encompass_Preferred
Bz-rA Phosphoramidite, also known as DMT-2'O-TBDMS-rA(bz) Phosphoramidite, is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection. It is used in oligonucleotide synthesis and intended for research use only.
  • A 2'-O-protected ribonucleoside phosphoramidite
  • Used in oligonucleotide synthesis
  • Suitable for laboratory chemicals and manufacture of substances
  • Can be used in the synthesis of 2'-O-guanidinopropyl-modified nucleoside phosphoramidites
  • For research use only and for experienced personnel
Catalog No. 50-004-46600 Supplier Medchemexpress LLC Supplier No. HYW00610210MM/1ML
Bz-rA Phosphoramidite, also known as DMT-2'O-TBDMS-rA(bz) Phosphoramidite, is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection. It is used in oligonucleotide synthesis and intended for research use only.
  • A 2'-O-protected ribonucleoside phosphoramidite
  • Used in oligonucleotide synthesis
  • Suitable for laboratory chemicals and manufacture of substances
  • Can be used in the synthesis of 2'-O-guanidinopropyl-modified nucleoside phosphoramidites
  • For research use only and for experienced personnel