Learn More
Bachem Bz-Arg-Gly-Phe-Phe-Pro-4MbNA · HCl
SDP
Catalog No. 501947023
Encompass_Preferred
Bz-Arg-Gly-Phe-Phe-Pro-4MbNA · HCl, 201928-98-5, C49H55N9O7 · HCl, Mr 918.49, 50mg. Bz-RGFFP-MNA is used for the colorimetric or fluorometric determination of cathepsin D. Excitation at ca. 335 nm, emission at ca. 410 nm.
Catalog No. 50-194-7023 Supplier Bachem Supplier No. 4002124.0050
Add to Cart
Add to Cart
Bz-Arg-Gly-Phe-Phe-Pro-4MbNA · HCl, 201928-98-5, C49H55N9O7 · HCl, Mr 918.49, 50mg. Bz-RGFFP-MNA is used for the colorimetric or fluorometric determination of cathepsin D. Excitation at ca. 335 nm, emission at ca. 410 nm.