Learn More
Cayman Chemical Bz-RS-ISer 3-Ph-OMe 5g
SDP
Catalog No. 502942524
Encompass_Preferred
An intermediate in the synthesis of 7-epi-10-deacetyltaxol
Catalog No. 50-294-2524 Supplier Cayman Chemical Supplier No. 402915 G
An intermediate in the synthesis of 7-epi-10-deacetyltaxol