Learn More
Carbosynth LLC. Bz-Tyr-pNA, 0.1g, MW: 405.4, Molecular formula: C22H19N3O5, CAS# 6154-45-6
SDP
Supplier:  Carbosynth LLC. SYN30150.1G
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements
Synonyms: Benzoyl-L-Tyrosine p-Nitroanilide, Purity: , Grade A, Application: Substrate for Chymotrypsin, Storage Temperature: -20C, References: G.A. Grant, A.Z. Eisen, and R.A. Bradshaw, In, Proteolytic Enzymes Part C, Methods in Enzymology, Vol. 80, (L. Lorand, Ed.), Academic Press, New York, 1981, pp. 722-734.
Catalog No. 50-168-7180