Learn More
Cayman Chemical BzNleKRRAMChydrochlorIde 10mg
SDP
Catalog No. 502940003
Encompass_Preferred
A fluorogenic substrate for YFV NS3 or DV NS2B/3 serine proteases; AMC is released upon enzymatic cleavage by YFV NS3 or DV NS2B/3 serine proteases and its fluorescence can be used to quantify YFV NS3 or DV NS2B/3 serine protease activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-294-0003 Supplier Cayman Chemical Supplier No. 2771010 MG
A fluorogenic substrate for YFV NS3 or DV NS2B/3 serine proteases; AMC is released upon enzymatic cleavage by YFV NS3 or DV NS2B/3 serine proteases and its fluorescence can be used to quantify YFV NS3 or DV NS2B/3 serine protease activity; ex/em = 340-360/440-460 nm, respectively