Learn More
Cayman Chemical BZO-HEPOXIZID 1mg
SDP
Catalog No. 502919321
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-291-9321 Supplier Cayman Chemical Supplier No. 369461 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications