Learn More
Cayman Chemical BZO-POXIZID-d9 1mg
SDP
Catalog No. 502913275
Encompass_Preferred
An internal standard for the quantification of BZO-POXIZID by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-291-3275 Supplier Cayman Chemical Supplier No. 367991 MG
An internal standard for the quantification of BZO-POXIZID by GC- or LC-MS; intended for research and forensic applications