Learn More
Aviva Systems Biology Corp BZW1 Antibody - N-terminal region (ARP62611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BZW1. It was validated on Western Blot. . Alternate names: BZAP45, KIAA0005, Nbla10236ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-723