Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-021 | 864289-85-0 | 100.0% | 467.65 g/mol | C27H41N5O2 | 25 MG
SDP

Catalog No. 5000244922
Encompass_Preferred

C-021 is a small-molecule CCR4 antagonist supplied for research use. It inhibits CCR4-mediated chemotaxis in human and mouse cells and blocks CCL22-induced GTPγS binding, making it suitable for biochemical and cellular assays.

  • Target: CCR4 (GPCR), suitable for immunology and GPCR research.
  • Potent CCR4 antagonist with reported IC50s of 140 nM (human chemotaxis), 39 nM (mouse chemotaxis), and 18 nM in GTPγS binding assays.
  • Molecular formula C27H41N5O2 and molecular weight 467.65 g/mol.
  • Reported purity 99.95% and solid appearance (off-white to light yellow).
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or animal therapeutic use.

Catalog No. 50-002-44922 Supplier Medchemexpress LLC Supplier No. HY10336425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-021 is a small-molecule CCR4 antagonist supplied for research use. It inhibits CCR4-mediated chemotaxis in human and mouse cells and blocks CCL22-induced GTPγS binding, making it suitable for biochemical and cellular assays.

  • Target: CCR4 (GPCR), suitable for immunology and GPCR research.
  • Potent CCR4 antagonist with reported IC50s of 140 nM (human chemotaxis), 39 nM (mouse chemotaxis), and 18 nM in GTPγS binding assays.
  • Molecular formula C27H41N5O2 and molecular weight 467.65 g/mol.
  • Reported purity 99.95% and solid appearance (off-white to light yellow).
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.