Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-021 | 864289-85-0 | 100.0% | 467.65 g/mol | C27H41N5O2 | 5 MG
SDP

Catalog No. 5000244538
Encompass_Preferred

C-021 is a small-molecule antagonist of the CC chemokine receptor 4 (CCR4) used for research into GPCR signaling and immunology. It inhibits CCR4-mediated chemotaxis in human and mouse systems and is supplied as a solid for laboratory studies.

  • Target CCR4.
  • Molecular formula C27H41N5O2.
  • Molecular weight 467.65 g/mol.
  • Purity 99.95%.
  • Appearance solid, off-white to light yellow.
  • Storage powder: -20°C (up to 3 years); in solvent: -80°C (short-term).
  • Supplied in small research quantities.

Catalog No. 50-002-44538 Supplier Medchemexpress LLC Supplier No. HY1033645MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-021 is a small-molecule antagonist of the CC chemokine receptor 4 (CCR4) used for research into GPCR signaling and immunology. It inhibits CCR4-mediated chemotaxis in human and mouse systems and is supplied as a solid for laboratory studies.

  • Target CCR4.
  • Molecular formula C27H41N5O2.
  • Molecular weight 467.65 g/mol.
  • Purity 99.95%.
  • Appearance solid, off-white to light yellow.
  • Storage powder: -20°C (up to 3 years); in solvent: -80°C (short-term).
  • Supplied in small research quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.