Learn More
Medchemexpress LLC HY-103364A 1mg Medchemexpress, C-021 (dihydrochloride) CAS:1784252-84-1 Purity:>98%
SDP
Catalog No. 501968087
Encompass_Preferred
Medchemexpress, HY-103364A 1mg C-021 (diHCl) CAS:1784252-84-1 C-021 dihydrochloride is a potent CC chemokine receptor-4 (CCR4) antagonist. C-021 dihydrochloride potently inhibits functional chemotaxis in human and mouse with IC50s of 140 nM and 39 nM, respectively. C-021 effectively prevents human CCL22-derived [35S]GTPγS from binding to the receptor with an IC50 of 18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8087 Supplier Medchemexpress LLC Supplier No. HY103364A1MG
Medchemexpress, HY-103364A 1mg C-021 (diHCl) CAS:1784252-84-1 C-021 dihydrochloride is a potent CC chemokine receptor-4 (CCR4) antagonist. C-021 dihydrochloride potently inhibits functional chemotaxis in human and mouse with IC50s of 140 nM and 39 nM, respectively. C-021 effectively prevents human CCL22-derived [35S]GTPγS from binding to the receptor with an IC50 of 18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.