Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-021 dihydrochloride | 1784252-84-1 | 99.9% | 540.57 g/mol | C27H43Cl2N5O2 | 10 MG
SDP

Catalog No. 5000244603
Encompass_Preferred

C-021 dihydrochloride is the dihydrochloride salt of a small-molecule antagonist of the CC chemokine receptor 4 (CCR4) provided for laboratory research. It inhibits CCR4-mediated chemotaxis in human and mouse systems and has been characterized in functional and binding assays.

  • Potent CC chemokine receptor 4 (CCR4) antagonist with reported low-nanomolar activity.
  • High chemical purity (reported ~99.9%).
  • Supplied as a solid; common pack sizes include 10 mg.
  • Molecular formula C27H43Cl2N5O2; molecular weight 540.57 g/mol.
  • Intended for research and analytical applications only.

Catalog No. 50-002-44603 Supplier Medchemexpress LLC Supplier No. HY103364A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-021 dihydrochloride is the dihydrochloride salt of a small-molecule antagonist of the CC chemokine receptor 4 (CCR4) provided for laboratory research. It inhibits CCR4-mediated chemotaxis in human and mouse systems and has been characterized in functional and binding assays.

  • Potent CC chemokine receptor 4 (CCR4) antagonist with reported low-nanomolar activity.
  • High chemical purity (reported ~99.9%).
  • Supplied as a solid; common pack sizes include 10 mg.
  • Molecular formula C27H43Cl2N5O2; molecular weight 540.57 g/mol.
  • Intended for research and analytical applications only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.