Learn More
TARGETMOL CHEMICALS INC C-021 DIHYDROCHLORIDE 5MG
SDP
Catalog No. 502581698
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. C-021 dihydrochloride is a potent CCR4 antagonist. It potently inhibits functional chemotaxis in human and mouse with IC50s of 140 nM and 39 nM. purity: 98%
Catalog No. 50-258-1698 Supplier TARGETMOL CHEMICALS INC Supplier No. T106395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. C-021 dihydrochloride is a potent CCR4 antagonist. It potently inhibits functional chemotaxis in human and mouse with IC50s of 140 nM and 39 nM. purity: 98%