Learn More
Sigma Aldrich c-(2-m-tolyl-thiazol-4-yl)-methylamine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Kinetics and mechanism of gas-phase elimination of trimethyl orthobutyrate has been examined over the temperature range of 310-369c and pressure range of 50-130torr.trimethyl orthobutyrate has been used in the preparation of:
- 5-acetamido-9-o-butyroyl-3,5-dideoxy-d-glycero-2-nonulo pyranosonic acid
- 9-o-butyroyl-3,5-dideoxy-5-glycoloylamido-d-glycero-2-nonulo pyranosonic acid
Specifications
Specifications
| Molecular Formula | C11H12N2S |
| Linear Formula | C11H12N2S |
| MDL Number | MFCD04055034 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 204.29 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.