Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-390, G-Biosciences
SCI_ED

Catalog No. S139091
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139091 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139091 Supplier G-Biosciences Supplier No. RC904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-390 is an antibacterial agent which is used for selective isolation of Pseudomonas aeruginosa from other bacterial species.

C-390 is primarily used in culture media to differentiate P. aeruginosa from other bacteria, especially within the Pseudomonas genus and other non-fermenters

Chemical Identifiers

Molecular Weight (g/mol) 490.05

Specifications

Physical Form Powder
Quantity 250 mg
Product Line Gibco
Boiling Point 546.6°C
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.