Learn More
Essentra Porous Technologies FG Clean Wipes 6-C43, Pre-Saturated, Ultra-Low Linting, 600 Wipes/Case
SDP
Catalog No. NC0103915
Encompass_Preferred
FG Clean Wipes 6-C43 are pre-saturated ultra-low linting wipes infused with 96% IPA for effective cleaning in controlled environments. Designed for critical cleaning and maintenance, these durable wipes are individually wrapped for convenience and quick use. Their superior oil and residue removal coupled with abrasion resistance make them ideal for scientific and industrial applications, setting them apart from other cleaning solutions.
This item is not returnable. View return policy
Catalog No. NC0103915 Supplier Essentra Porous Technologies Supplier No. 6C43
FG Clean Wipes 6-C43 are pre-saturated ultra-low linting wipes infused with 96% IPA for effective cleaning in controlled environments. Designed for critical cleaning and maintenance, these durable wipes are individually wrapped for convenience and quick use. Their superior oil and residue removal coupled with abrasion resistance make them ideal for scientific and industrial applications, setting them apart from other cleaning solutions.