Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C 646, Tocris Bioscience™

Catalog No. 420050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-005-0 50 mg
42-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-005-0 Supplier R&D Systems Supplier No. 4200/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Selective p300/CBP inhibitor

Selective p300/CREB-binding protein (CBP) inhibitor (Ki = 400 nM). Selective for p300 over six other histone acetyltransferases (HATs). Suppresses histone H3 and H4 acetylation in mouse fibroblast cell lines.

This compound is a mixture of Z:E isomers (∽ 80:20).

Chemical Identifiers

CAS 328968-36-1
Molecular Formula C24H19N3O6
Molecular Weight (g/mol) 445.43
MDL Number MFCD01784780
InChI Key HEKJYZZSCQBJGB-UHFFFAOYSA-N
Synonym 4-4-5-4,5-dimethyl-2-nitrophenyl furan-2-yl methylidene-3-methyl-5-oxopyrazol-1-yl benzoic acid
PubChem CID 2871948
IUPAC Name 4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid
SMILES CC1=NN(C(=O)C1=CC1=CC=C(O1)C1=CC(C)=C(C)C=C1[N+]([O-])=O)C1=CC=C(C=C1)C(O)=O

Specifications

Quantity 50 mg
Formula Weight 445.42
Percent Purity >98%
Chemical Name or Material C 646
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.