Learn More
Abcam C 7280948, PRMT1 inhibitor, 50MG
Catalog No. 502705489
Encompass_Preferred
MW 276.36 Da, Purity >99%. PRMT1 inhibitor (IC₅₀ = 12.75 μM). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5489 Supplier Abcam Supplier No. AB14482950MG
MW 276.36 Da, Purity >99%. PRMT1 inhibitor (IC₅₀ = 12.75 μM). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement