Learn More
Medchemexpress LLC C-82 | 1422253-37-9 | 99.9% | 578.66 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1123591MG
Encompass_Preferred
C-82 is a second-generation specific CBP/β-catenin antagonist. It functions by inhibiting the binding between β-catenin and CBP, while simultaneously increasing the binding between β-catenin and p300. C-82 is the active form of PRI-724, which is rapidly hydrolyzed to C-82 in vivo and has shown an acceptable toxicity profile in preclinical studies.
  • Inhibits CBP/β-catenin binding.
  • Increases β-catenin/p300 binding.
  • Inhibits activation of quiescent hepatic stellate cells (HSCs).
  • Promotes cell death in culture-activated HSCs.
Catalog No. 50-004-36429