Learn More
Medchemexpress LLC HY-100735 5mg Medchemexpress, C 87 CAS:332420-90-3 Purity:>98%
SDP
Catalog No. 501931931
Encompass_Preferred
Medchemexpress, HY-100735 5mg C 87 CAS:332420-90-3 C87 is a novel small-molecule TNFα inhibitor; potently inhibits TNFα-induced cytotoxicity with an IC 50 of 8.73 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1931 Supplier Medchemexpress LLC Supplier No. HY1007355MG
Medchemexpress, HY-100735 5mg C 87 CAS:332420-90-3 C87 is a novel small-molecule TNFα inhibitor; potently inhibits TNFα-induced cytotoxicity with an IC 50 of 8.73 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.