Learn More
Bachem (Cys18)-Atrial Natriuretic Factor (4-18) amide (mouse, rabbit, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4030384.0500
Encompass_Preferred
(Cys18)-Atrial Natriuretic Factor (4-18) amide (mouse, rabbit, rat) Trifluoroacetate, H-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Cys-NH2, 111863-73-1, C64H107N25O19S2, Mr 1594.84, 0.5mg. C-ANF (4-23), (Des-Gln18,des-Ser19,des-Gly20·22,des-Leu21)-Atrial Natriuretic Peptide (4-23) amide (mouse, rabbit, rat), Des[Gln18, Ser19, Gly20,22, Leu21] ANP (4-23), amide, rat. C-ANF 4-23 binds with high affinity to approximately 99% of ANF receptors in the isolated perfused rat kidney. It competes effectively with biologically active atrial peptides for binding sites but is devoid of agonist or antagonist action on the generation of cGMP in vascular smooth muscle and endothelial cells in culture.
Catalog No. 50-194-6762