Learn More
Medchemexpress LLC c(avb6)-DOTA TFA | 98.6% | 1546.65 | 1 MG
SDP
Catalog No. 5000397831
Encompass_Preferred
c(avb6)-DOTA TFA is a conjugate of the αvβ6 integrin selective peptide ring modified with the DOTA chelator. The Lu(III) complex of c(avb6)-DOTA TFA shows a comparable affinity for αvβ6 integrin (IC50=0.8 nM) and can be utilized as a tumor diagnostic.
  • Conjugate of αvβ6 integrin selective peptide ring
  • Modified with DOTA chelator
  • Lu(III) complex shows comparable affinity for αvβ6 integrin (IC50=0.8 nM)
  • Can be utilized as a tumor diagnostic
  • Intended for research use only
Catalog No. 50-003-97831 Supplier Medchemexpress LLC Supplier No. HYP63511MG
c(avb6)-DOTA TFA is a conjugate of the αvβ6 integrin selective peptide ring modified with the DOTA chelator. The Lu(III) complex of c(avb6)-DOTA TFA shows a comparable affinity for αvβ6 integrin (IC50=0.8 nM) and can be utilized as a tumor diagnostic.
  • Conjugate of αvβ6 integrin selective peptide ring
  • Modified with DOTA chelator
  • Lu(III) complex shows comparable affinity for αvβ6 integrin (IC50=0.8 nM)
  • Can be utilized as a tumor diagnostic
  • Intended for research use only