Learn More
PATTERSON DENTAL COMPANY C&B-Metabond Quick Base, Bottle (10 ml)
Catalog No. NC1013606
Encompass
C&B-METABOND adheres to most surfaces you encounter in dentistry (cast metal, amalgam, dentin, enamel, cementum, acrylic or composite). Use the system to make fast intraoral ceramic repairs. C&B-METABOND chemically grafts to any composite.
Catalog No. NC1013606 Supplier PATTERSON DENTAL COMPANY Supplier No. 075533492
C&B-METABOND adheres to most surfaces you encounter in dentistry (cast metal, amalgam, dentin, enamel, cementum, acrylic or composite). Use the system to make fast intraoral ceramic repairs. C&B-METABOND chemically grafts to any composite.