Learn More
Cusabio Technology LLC C-C motif chemokine 4
Catalog No. 5000048904
Encompass_Preferred
This kit detects C-C motif chemokine 4 (CCL4) in human serum plasma and tissue homogenates providing a reliable quantitative analysis through a sandwich ELISA method It features a detection range of 31 2 pg/ml to 2000 pg/ml with a sensitivity as low as 7 8 pg/ml ensuring precise measurement across biological samples The assay can be completed within 1-5 hours requiring only 50-100 L of sample per test Absorbance is measured at 450 nm to determine CCL4 concentrations making it suitable for research applications requiring accurate cytokine profiling The kit s standardized protocol and consistent performance support efficient data generation in immunology and inflammation studies
Catalog No. 50-000-48904 Supplier Cusabio Technology LLC Supplier No. CSBEL004797HU
This kit detects C-C motif chemokine 4 (CCL4) in human serum plasma and tissue homogenates providing a reliable quantitative analysis through a sandwich ELISA method It features a detection range of 31 2 pg/ml to 2000 pg/ml with a sensitivity as low as 7 8 pg/ml ensuring precise measurement across biological samples The assay can be completed within 1-5 hours requiring only 50-100 L of sample per test Absorbance is measured at 450 nm to determine CCL4 concentrations making it suitable for research applications requiring accurate cytokine profiling The kit s standardized protocol and consistent performance support efficient data generation in immunology and inflammation studies