Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-C motif chemokine 5 (RANTES) | 10 UG
SDP

Catalog No. 5000293266
Encompass_Preferred

Recombinant rat RANTES/CCL5 is a biologically active chemokine produced in Pichia pastoris and supplied as a filtered PBS solution with a His tag for research use in in vitro assays and chemotaxis studies.

  • Expressed in Pichia pastoris for eukaryotic post-translational processing.
  • His-tagged recombinant protein for simplified detection and purification.
  • Sequence corresponds to residues S25-S92 (accession P50231).
  • Endotoxin level <1 EU/μg, suitable for cell-based assays.
  • Biological activity: ED50 = 8.964 ng/mL in a CCR5 chemotaxis assay.
  • Formulated in 0.22 μm filtered PBS, pH 7.4.
  • Store at -80°C for long term; ship with dry ice; avoid repeated freeze-thaw.

Catalog No. 50-002-93266 Supplier Medchemexpress LLC Supplier No. HYP7460210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat RANTES/CCL5 is a biologically active chemokine produced in Pichia pastoris and supplied as a filtered PBS solution with a His tag for research use in in vitro assays and chemotaxis studies.

  • Expressed in Pichia pastoris for eukaryotic post-translational processing.
  • His-tagged recombinant protein for simplified detection and purification.
  • Sequence corresponds to residues S25-S92 (accession P50231).
  • Endotoxin level <1 EU/μg, suitable for cell-based assays.
  • Biological activity: ED50 = 8.964 ng/mL in a CCR5 chemotaxis assay.
  • Formulated in 0.22 μm filtered PBS, pH 7.4.
  • Store at -80°C for long term; ship with dry ice; avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.