Learn More
Electron Microscopy Sciences C-Chip Disposable Hemacytometer, Neubauer Improved, 200 tests, 50 slides
SDP
Catalog No. NC1713722
Encompass_Preferred
The world's first truly disposable Hemacytometer, the C-Chip offers precise volume control and is easy to use.
Catalog No. NC1713722 Supplier Electron Microscopy Sciences Supplier No. 6350802
The world's first truly disposable Hemacytometer, the C-Chip offers precise volume control and is easy to use.